Order Entry
Italy
ContactUsLinkComponent
Soluble TNF-related apoptosis-inducing ligand Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain c ontaining 169 amino acids (114-281) and having a molecular mass of 19.6 kDa. The sTRAIL is purified by propr 1 * 10 µG
Catalog # BIRBORB80630-10
Supplier:  Biorbyt
Soluble TNF-related apoptosis-inducing ligand Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain c ontaining 169 amino acids (114-281) and having a molecular mass of 19.6 kDa. The sTRAIL is purified by propr 1 * 10 µG
Catalog # BIRBORB80630-10
Supplier:  Biorbyt