Recombinant Human Cu/Zn Superoxide Dismutase produced in E.coli is homodimer, non-glycosylated polypeptide chain containing 2x 154 a mino acids and having total molecular mass of 31,608 Dalton. The Cu/Zn SOD is purified by proprietary chroma 1 * 100 µG
Catalog # BIRBORB80178-100
Supplier: Biorbyt
Frequently Bought Together