Order Entry
Italy
ContactUsLinkComponent
TNF Receptor Human Recombinant produced in CHO is dimeric, glycosylated, polypeptide chain consisting of the extracellular ligand-bi nding portion of the human 75 kilodalton (p75) TNF receptor (TNFR) linked to the Fc portion of human IgG1. T 1 * 50 µG
Catalog # BIRBORB80622-50
Supplier:  Biorbyt
TNF Receptor Human Recombinant produced in CHO is dimeric, glycosylated, polypeptide chain consisting of the extracellular ligand-bi nding portion of the human 75 kilodalton (p75) TNF receptor (TNFR) linked to the Fc portion of human IgG1. T 1 * 50 µG
Catalog # BIRBORB80622-50
Supplier:  Biorbyt