Order Entry
Italy
ContactUsLinkComponent
Flavokinase Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 182 amino acids (1-162 a.a.) and having a molecular mass of 20.5kDa. Flavokinase is fused to 20 a.a. His-Tag at N-terminus and puri 1 * 20 µG
Catalog # BIRBORB80807-20
Supplier:  Biorbyt
Flavokinase Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 182 amino acids (1-162 a.a.) and having a molecular mass of 20.5kDa. Flavokinase is fused to 20 a.a. His-Tag at N-terminus and puri 1 * 20 µG
Catalog # BIRBORB80807-20
Supplier:  Biorbyt