Order Entry
Italy
ContactUsLinkComponent
TGF-b Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 112 amino acids fragment (279 -390) having molecular weight of 17.3kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The TGF-b H 1 * 10 µG
Catalog # BIRBORB80610-10
Supplier:  Biorbyt
TGF-b Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 112 amino acids fragment (279 -390) having molecular weight of 17.3kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The TGF-b H 1 * 10 µG
Catalog # BIRBORB80610-10
Supplier:  Biorbyt