Order Entry
Italy
ContactUsLinkComponent
DHH Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 197 amino acids (23-198) and ha ving a molecular mass of 22 kDa. DHH is fused to His-tag (20 a.a.) at N-terminus and is purified by propriet 1 * 5 µG
Catalog # BIRBORB80582-5
Supplier:  Biorbyt
DHH Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 197 amino acids (23-198) and ha ving a molecular mass of 22 kDa. DHH is fused to His-tag (20 a.a.) at N-terminus and is purified by propriet 1 * 5 µG
Catalog # BIRBORB80582-5
Supplier:  Biorbyt