Order Entry
Italy
ContactUsLinkComponent
HGF-A Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 463 amino acids fragment (32- 494) having total molecular weight of 57.8kDa. The HGF-A is fused with 4.5kDa amino-terminal hexahistidine t 1 * 10 µG
Catalog # BIRBORB80381-10
Supplier:  Biorbyt
HGF-A Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 463 amino acids fragment (32- 494) having total molecular weight of 57.8kDa. The HGF-A is fused with 4.5kDa amino-terminal hexahistidine t 1 * 10 µG
Catalog # BIRBORB80381-10
Supplier:  Biorbyt