Order Entry
Italy
ContactUsLinkComponent
GMF-Gamma (GMF-Gamma) Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 142 amino acids and having total molecular mass of 16.8 kDa. GMF-Gamma, GMF-Gamma, Human Recombinant is purified by proprie 1 * 5 µG
Catalog # BIRBORB80377-5
Supplier:  Biorbyt
GMF-Gamma (GMF-Gamma) Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 142 amino acids and having total molecular mass of 16.8 kDa. GMF-Gamma, GMF-Gamma, Human Recombinant is purified by proprie 1 * 5 µG
Catalog # BIRBORB80377-5
Supplier:  Biorbyt