Order Entry
Italy
ContactUsLinkComponent
Prolactin-His Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids fragme nt (29-227) and having a molecular mass of 23 kDa with an amino-terminal hexahistidine tag. The Prolactin is 1 * 20 µG
Codice catalogo BIRBORB80559-20
Supplier:  Biorbyt
Prolactin-His Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids fragme nt (29-227) and having a molecular mass of 23 kDa with an amino-terminal hexahistidine tag. The Prolactin is 1 * 20 µG
Codice catalogo BIRBORB80559-20
Supplier:  Biorbyt