Order Entry
Italy
ContactUsLinkComponent
CDK2AP1 Human Recombinant fused to 37 amino acid His Tag at N-terminal produced in E.coli is a single, non-glycosylated, polypeptide chain containing 152 amino acids (1-115 a.a.) and having a molecular mass of 16.6 kDa. The CDK2AP1 is purif 1 * 5 µG
Catalog # BIRBORB80778-5
Supplier:  Biorbyt
CDK2AP1 Human Recombinant fused to 37 amino acid His Tag at N-terminal produced in E.coli is a single, non-glycosylated, polypeptide chain containing 152 amino acids (1-115 a.a.) and having a molecular mass of 16.6 kDa. The CDK2AP1 is purif 1 * 5 µG
Catalog # BIRBORB80778-5
Supplier:  Biorbyt