Order Entry
Italy
ContactUsLinkComponent
CCNH Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 343 amino acids (1-323 a.a.) a nd having a molecular mass of 39.8 kDa. The CCNH is fused to 20 amino acid His Tag and purified by proprieta 1 * 5 µG
Catalog # BIRBORB80777-5
Supplier:  Biorbyt
CCNH Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 343 amino acids (1-323 a.a.) a nd having a molecular mass of 39.8 kDa. The CCNH is fused to 20 amino acid His Tag and purified by proprieta 1 * 5 µG
Catalog # BIRBORB80777-5
Supplier:  Biorbyt