Order Entry
Italy
ContactUsLinkComponent
AURKB Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 364 amino acids (1-344) and h aving a molecular mass of 41.4kDa. AURKB is fused to 20 a.a. His-Tag at N-terminus and purified by proprieta 1 * 10 µG
Catalog # BIRBORB80775-10
Supplier:  Biorbyt
AURKB Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 364 amino acids (1-344) and h aving a molecular mass of 41.4kDa. AURKB is fused to 20 a.a. His-Tag at N-terminus and purified by proprieta 1 * 10 µG
Catalog # BIRBORB80775-10
Supplier:  Biorbyt