Order Entry
Italy
ContactUsLinkComponent
Progranulin Human Recombinant fused to FLAG at C-terminus produced in HEK is a single, glycosylated, polypeptide chain containing 59 3 amino acids and having a molecular mass of 88 kDa. The Progranulin is purified by standard chromatographic 1 * 10 µG
Catalog # BIRBORB80550-10
Supplier:  Biorbyt
Progranulin Human Recombinant fused to FLAG at C-terminus produced in HEK is a single, glycosylated, polypeptide chain containing 59 3 amino acids and having a molecular mass of 88 kDa. The Progranulin is purified by standard chromatographic 1 * 10 µG
Catalog # BIRBORB80550-10
Supplier:  Biorbyt