Order Entry
Italy
ContactUsLinkComponent
GDF5 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 141 amino acids (382-501 a.a.) and having total molecular mass of 15.8 kDa. GDF5 is fused to 20 amino acid His Tag at N-terminus and purif 1 * 5 µG
Catalog # BIRBORB80339-5
Supplier:  Biorbyt
GDF5 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 141 amino acids (382-501 a.a.) and having total molecular mass of 15.8 kDa. GDF5 is fused to 20 amino acid His Tag at N-terminus and purif 1 * 5 µG
Catalog # BIRBORB80339-5
Supplier:  Biorbyt