Order Entry
Italy
ContactUsLinkComponent
MTPN Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 138 amino acids (1-118 a.a.) an d having a molecular mass of 15 kDa. The MTPN is fused to 20 amino acid His-Tag at N-terminus and purified b 1 * 5 µG
Codice catalogo BIRBORB80525-5
Supplier:  Biorbyt
MTPN Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 138 amino acids (1-118 a.a.) an d having a molecular mass of 15 kDa. The MTPN is fused to 20 amino acid His-Tag at N-terminus and purified b 1 * 5 µG
Codice catalogo BIRBORB80525-5
Supplier:  Biorbyt