Order Entry
Italy
ContactUsLinkComponent
MCSF Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 179 amino acids (33-190 a.a.) and having total molecular mass of 20.7 kDa.MCSF is fused to 20 amino acid His Tag at N-terminus and purifie 1 * 25 µG
Catalog # BIRBORB80518-25
Supplier:  Biorbyt
MCSF Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 179 amino acids (33-190 a.a.) and having total molecular mass of 20.7 kDa.MCSF is fused to 20 amino acid His Tag at N-terminus and purifie 1 * 25 µG
Catalog # BIRBORB80518-25
Supplier:  Biorbyt