Order Entry
Italy
ContactUsLinkComponent
Fibroblast Growth Factor -21 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 183 am ino acids including N-terminal Methionin and having a molecular mass of 20.1 kDa. The amino acid sequence of 1 * 10 µG
Catalog # BIRBORB80319-10
Supplier:  Biorbyt
Fibroblast Growth Factor -21 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 183 am ino acids including N-terminal Methionin and having a molecular mass of 20.1 kDa. The amino acid sequence of 1 * 10 µG
Catalog # BIRBORB80319-10
Supplier:  Biorbyt