Order Entry
Italy
ContactUsLinkComponent
Recombinant Rat CT-1 is a member of the IL-6 family of cytokines which also includes LIF, CNTF, OSM (Oncostatin M), IL-11, IL-6 and possibly NNT-1/ BCSF-3. CT-1 is a pleiotropic cytokine which is expressed in various tissues including the a 1 * 10 µG
Catalog # BIRBORB107701-10
Supplier:  Biorbyt
Recombinant Rat CT-1 is a member of the IL-6 family of cytokines which also includes LIF, CNTF, OSM (Oncostatin M), IL-11, IL-6 and possibly NNT-1/ BCSF-3. CT-1 is a pleiotropic cytokine which is expressed in various tissues including the a 1 * 10 µG
Catalog # BIRBORB107701-10
Supplier:  Biorbyt