Order Entry
Italy
ContactUsLinkComponent
Defensin are 3-4 kDa antimicrobial peptides of which three distinct families have been identified alpha -defensin, beta -defensins, andinsect defensins. Mouse beta -Defensin 1, as a member of the beta -defensin family, is a salt-sensitive 1 * 20 µG
Catalog # BIRBORB107687-20
Supplier:  Biorbyt
Defensin are 3-4 kDa antimicrobial peptides of which three distinct families have been identified alpha -defensin, beta -defensins, andinsect defensins. Mouse beta -Defensin 1, as a member of the beta -defensin family, is a salt-sensitive 1 * 20 µG
Catalog # BIRBORB107687-20
Supplier:  Biorbyt