Order Entry
Italy
ContactUsLinkComponent
Human Interleukin-16 (IL-16) is also called lymphocyte chemoattractant factor (LCF) and it has three isoforms which arecleaved from pre-Interleukin-16. IL-16 is mostly secreted by lymphocytes, epithelial cells, eosinophils, and CD8+ T cells 1 * 2 µG
Catalog # BIRBORB107756-2
Supplier:  Biorbyt
Human Interleukin-16 (IL-16) is also called lymphocyte chemoattractant factor (LCF) and it has three isoforms which arecleaved from pre-Interleukin-16. IL-16 is mostly secreted by lymphocytes, epithelial cells, eosinophils, and CD8+ T cells 1 * 2 µG
Catalog # BIRBORB107756-2
Supplier:  Biorbyt