Order Entry
Italy
ContactUsLinkComponent
Vascular Endothelial Growth Factor is a potent growth and angiogenic cytokine. It stimulates proliferation and survival ofendothelia l cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays 1 * 10 µG
Catalog # BIRBORB107815-10
Supplier:  Biorbyt
Vascular Endothelial Growth Factor is a potent growth and angiogenic cytokine. It stimulates proliferation and survival ofendothelia l cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays 1 * 10 µG
Catalog # BIRBORB107815-10
Supplier:  Biorbyt