Dish-shaped Dewar flasks, type SCH-CAL
Dewar lid for SCH 20 CAL vessel, Dewar flask SCH 20 CAL
Catalog # KGWA1674
Supplier: KGW
Jump to:
Product Details & Documents
Silvered borosilicate glass, structured aluminium protective covering. For tempering of round bottom flasks, cooling of small samples. They are vacuum insulated and silvered.
- Suitable as hot or cold bath
- Can be used on a magnetic stirrer
- The cover is anti-magnetic and stainless
Temperature range from −200 to +200 °C.
CAL flasks have an external stucco aluminium protective casing, alternatively the glass inserts (without casing) are also available.
Specifications
- Description:Dewar lid for SCH 20 CAL vessel
- Seq:12
- For:Dewar flask SCH 20 CAL