Order Entry
Italy
ContactUsLinkComponent
C18Q x 3µm particle size x 100Å pore size, 100mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-03M46P
C18Q x 3µm particle size x 100Å pore size, 100mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-03M46P
Frequently Bought Together