Order Entry
Italy
ContactUsLinkComponent
PFP x 3µm particle size x 120Å pore size, 150mm length x 3.0mm ID, Stainless steel with guard ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2PFP-03N03S
undefined
PFP x 3µm particle size x 120Å pore size, 150mm length x 3.0mm ID, Stainless steel with guard ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2PFP-03N03S
Frequently Bought Together