Order Entry
Italy
ContactUsLinkComponent
Phenyl Hexylx 5µm particle size x 120Å pore size, 50mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2NH4-05L03S
Phenyl Hexylx 5µm particle size x 120Å pore size, 50mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2NH4-05L03S
Frequently Bought Together