Order Entry
Italy
ContactUsLinkComponent
HILIC HC x 5µm particle size x 120Å pore size, 250mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL9-05R46P
undefined
HILIC HC x 5µm particle size x 120Å pore size, 250mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL9-05R46P
Frequently Bought Together