Order Entry
Italy
ContactUsLinkComponent
C8H x 5µm particle size x 120Å pore size, 150mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C83-05N30K
C8H x 5µm particle size x 120Å pore size, 150mm length x 300µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C83-05N30K
Frequently Bought Together