Order Entry
Italy
ContactUsLinkComponent
TRAY-CHIP 2 H20-130-16WH-62C02 Material: STAT-PRO 150 (Black Conductive Polypropylene) Pocket Dimensions: 3.30mm x 3.30mm x 0.41mm Capacity: 10x10 (100) 1 * 100 ST
Codice catalogo ENTEH2013016WH62C2
Supplier:  ENTEGRIS
undefined
TRAY-CHIP 2 H20-130-16WH-62C02 Material: STAT-PRO 150 (Black Conductive Polypropylene) Pocket Dimensions: 3.30mm x 3.30mm x 0.41mm Capacity: 10x10 (100) 1 * 100 ST
Codice catalogo ENTEH2013016WH62C2
Supplier:  ENTEGRIS
Frequently Bought Together