0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
Wipe Compo 7408 11X12 Polypro/Cellulose Material: Polypropylene / Cellulose Size 11´ x 12´ (27,94x30,48cm), Weight: 88gr / m² 1 unit = 100 wipes 14 bags x 100 = 1 carton 1 * 1.400 items
EPTA7-7408-ABL-00
:
ESSENTRA POROUS TECHNOLOGIES
Wipe Compo 7408 11X12 Polypro/Cellulose Material: Polypropylene / Cellulose Size 11´ x 12´ (27,94x30,48cm), Weight: 88gr / m² 1 unit = 100 wipes 14 bags x 100 = 1 carton 1 * 1.400 items
EPTA7-7408-ABL-00
:
ESSENTRA POROUS TECHNOLOGIES
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
pH indicator strips, MQuant®, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals