Order Entry
Italy
ContactUsLinkComponent
HRSU10 Barriere bags 130 x 110mm Material: complex film laminated PET 12µm / 12µm Alu / PE 75µm Vapor barrier: MVTR 0.05 g / m² / day (according to ASTM D 398-83) Barrier to oxygen: OTR 0.03 cm3 / m² / day (according to ASTM D 1434) Barrier to light: 1 * 2.500 items
Catalog # ATMIHRSU1015190002
Supplier:  PALL LIFESCIENCES
HRSU10 Barriere bags 130 x 110mm Material: complex film laminated PET 12µm / 12µm Alu / PE 75µm Vapor barrier: MVTR 0.05 g / m² / day (according to ASTM D 398-83) Barrier to oxygen: OTR 0.03 cm3 / m² / day (according to ASTM D 1434) Barrier to light: 1 * 2.500 items
Catalog # ATMIHRSU1015190002
Supplier:  PALL LIFESCIENCES
Frequently Bought Together