Order Entry
Italy
ContactUsLinkComponent
suitable for ISO class 5-6 cleanrooms, constructed from 100% polyester filament, cleanroom laundred cut edge, basis weight approx. 155g/m² ***bulk packed*** 1 * 150 items
Catalog # TEXWTX1009B_U
Supplier:  Texwipe
suitable for ISO class 5-6 cleanrooms, constructed from 100% polyester filament, cleanroom laundred cut edge, basis weight approx. 155g/m² ***bulk packed*** 1 * 150 items
Catalog # TEXWTX1009B_U
Supplier:  Texwipe
Frequently Bought Together