Wall-mounted cabinets
Wall- mounted cabinet, with one shelf, open front, -, 1200×350×480 mm
KOTT307-00014
Made of epoxy powder coated steel, with one shelf.
- Solid hinged doors or sliding glass doors
- Height adjustable shelfs
- Maximum loading capacity: 80 kg
- Description:Wall- mounted cabinet, with one shelf, open front
- Seq:14
- W×D×H:1200×350×480 mm
- Doors:-



