Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, lockable, 2 sliding glass doors, 1500×350×630 mm
  KOTT307-00027
Wall-mounted cabinets
  KOTT307-00027
Wall-mounted cabinets
Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, lockable, 2 sliding glass doors, 1500×350×630 mm

 
 
   
1 * 1 pezzi
 

Made of epoxy powder coated steel, with one shelf.

  • Solid hinged doors or sliding glass doors
  • Height adjustable shelfs
  • Maximum loading capacity: 80 kg
 
  • Description:
    Wall-mounted cabinet, with one shelf, lockable
  • Seq:
    29
  • W×D×H:
    1500×350×630 mm
  • Doors:
    2 sliding glass doors