Order Entry
Italy
Orders LinkContactUsLinkComponent
Wall-mounted cabinets

Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, lockable, 2 doors, 900×366×780 mm
Catalog # KOTT307-00075
Supplier:   KOETTERMANN
Wall-mounted cabinets
Catalog # KOTT307-00075
Supplier:  KOETTERMANN
Wall-mounted cabinets
Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, lockable, 2 doors, 900×366×780 mm

Price
Inquire For Price
Log in for availability
1 * 1 pezzi
Product Details & Documents

Made of epoxy powder coated steel, with one shelf.

  • Solid hinged doors or sliding glass doors
  • Height adjustable shelfs
  • Maximum loading capacity: 80 kg
Specifications
  • Description:
    Wall-mounted cabinet, with one shelf, lockable
  • Seq:
    12
  • W×D×H:
    900×366×780 mm
  • Doors:
    2 doors