Heating tapes, HST
Heating tape HST with 3 m length, 150 W output
Catalog # HORS025204
Supplier: HORST
Jump to:
Product Details & Documents
These slim heating tapes consist of two PTFE insulated heating conductors with earthed protection braid which have a common coating made of metal cloth.
Specifications
- Description:Heating tape HST with 3 m length, 150 W output
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



