Mildly alkaline special cleaning concentrate. For automated cleaning in laboratories. Free of chlorine and surfactants, liquid. Free of EDTA and NTA. Compatible also with parts made from aluminium.
Catalog # DECO500925.25-K10W
Supplier: Borer Chemie
Frequently Bought Together