Order Entry
Italy
ContactUsLinkComponent
pH combination electrode with Memosens screw plug head Liquid electrolyt comb. electrode with NTC 30K and plastic adapter for hose connection incl. Pg13.5 thread, platinum junction, electrolyte L 200, sphere membrane, A-glass, length 225 mm, 12 mm Ø 1 * 1 items
Catalog # SCOI285118185
Supplier:  SI Analytics
pH combination electrode with Memosens screw plug head Liquid electrolyt comb. electrode with NTC 30K and plastic adapter for hose connection incl. Pg13.5 thread, platinum junction, electrolyte L 200, sphere membrane, A-glass, length 225 mm, 12 mm Ø 1 * 1 items
Catalog # SCOI285118185
Supplier:  SI Analytics
Frequently Bought Together