Haematoxylin solution (Harris), Q Path® for microscopy
Bag-in-box, 5 L
Ready to use solution for Papanicolaou cytological staining.
: DcH-48, Natural Black I, Hydroxybrasilin
- :517-28-2
- :C₁₆H₁₄O₆
- :MFCD00078111
- :302.28 g/mol
- :Ambient
- Appearance:Clear liquid
- Colour of solution:Purplish red
- Density (20/4):1,053 - 1,059
- pH (20°C):2,2 - 2,6
- Absorbance (0.5 %):553 - 560 nm
- Pack type:Bag-in-box
- Pk:5 L



