Order Entry
Italy
Orders LinkContactUsLinkComponent
Formalin 10%, Q Path®, buffered

Formalin 10%, Q Path®, buffered

Bag-in-box, -, 10 L
Catalog # 11699408.
Health
Irritant
Supplier:   VWR Chemicals
Formalin 10%, Q Path®, buffered
Catalog # 11699408.
Supplier:  VWR Chemicals
CAS Number:  50-00-0
Formalin 10%, Q Path®, buffered
Formalin 10%, Q Path®, buffered

Bag-in-box, -, 10 L

CAS Number:  50-00-0
Price
76,20 €
Log in for availability
1 * 10 L
Product Details & Documents

Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.

Formaldehyde 4% is approximately equal to Formalin 10%.

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Density:1.013 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specification Test Results

  • Appearance:
    Colourless liquid
  • Test formaldehyde:
    Passes test
  • Density (20/4):
    1,012 - 1,014
  • pH (20°C):
    6,8 - 7,2

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Pk Info:
    -

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.