Order Entry
Italy
Orders LinkContactUsLinkComponent
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®

Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®

Phosphate Test, 0,015-0,03-0,045-0,06-0,08-0,11-0,14 mg/l PO₄-P, 0,046-0,092-0,14-0,18-0,25-0,34-0,43 mg/l PO₄, Phosphormolybdenum blue, MCC, 200 Tests
Codice catalogo 1.18394.0001
Nome del fornitore:   Merck
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®
Codice catalogo 1.18394.0001
Nome del fornitore:  Merck
Numero CAS:  
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®
Visual colour kits, phosphate, with colour card comparator, MQuant®, Supelco®

Phosphate Test, 0,015-0,03-0,045-0,06-0,08-0,11-0,14 mg/l PO₄-P, 0,046-0,092-0,14-0,18-0,25-0,34-0,43 mg/l PO₄, Phosphormolybdenum blue, MCC, 200 Tests

Numero CAS:  
Price
195,00 €
Log in for availability
1 * 200 Tests
Product Details & Documents

Quick tests, visual colour comparison.

  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available

MQuant® with colour card comparator

By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.

Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.

Specifications

Specifications

  • Chemical method:
    Phosphormolybdenum blue
  • Pk:
    200 Tests
  • Description:
    Phosphate Test
  • Type:
    MCC
  • Range:
    0,015-0,03-0,045-0,06-0,08-0,11-0,14 mg/l PO₄-P, 0,046-0,092-0,14-0,18-0,25-0,34-0,43 mg/l PO₄

Specifications