Order Entry
Italy
Orders LinkContactUsLinkComponent
 

High visibility rain jacket, Essential, class 3

Hi-vis yellow, 3XL
Catalog # KWIN1010121303XL
Supplier:   FRISTADS KANSAS
High visibility rain jacket, Essential, class 3
Catalog # KWIN1010121303XL
Supplier:  FRISTADS KANSAS
High visibility rain jacket, Essential, class 3
High visibility rain jacket, Essential, class 3

Hi-vis yellow, 3XL

Price
Inquire For Price
Log in for availability
1 * 1 pezzi
Product Details & Documents

Basic rain jacket made of polyurethane-coated nylon jersey with 'stretch' function.

  • Detachable drawstring hood
  • Buttons and zip
  • Adjustable cuffs

Weight: 170 g/m²

 : EN 471 (3-2), EN 343 (3-2)

Specifications
  • Clothing Size:
    3XL
  • Colour:
    Hi-vis yellow
  • Seq:
    08
  • Cat. No.:
    KWIN1010121303XL
  • Sleeve or leg length:
    Long
  • Risk area:
    Rain (waterproof)
  • Model:
    Class 3
  • Garment application:
    General Purpose
  • Gender:
    Unisex
  • Closure type:
    Zip closure
  • Fabric:
    Polyurethane-coated nylon
  • Weight:
    170 g/m²