Order Entry
Italy
Orders LinkContactUsLinkComponent
Disposable Post Mortem scalpels
Disposable Post Mortem scalpels
PM40, with standard blade
Catalog # SWAN2580
Supplier:   SWANN MORTON
Disposable Post Mortem scalpels
Catalog # SWAN2580
Supplier:  SWANN MORTON
Disposable Post Mortem scalpels
Disposable Post Mortem scalpels
PM40, with standard blade
Price
122,00 €
Log in for availability
1 * 5 pezzi
Product Details & Documents
Carbon blade. Non sterile, disposable histopathology and autopsy knives.
  • With easy to remove guard to protect the cutting edge: Users do not have to fix the blade to the handle
  • Available with either standard PM40 blade or PM40B bull nose blade fixed in a distinctive blue handle
Documents
Specifications
  • Type:
    PM40, with standard blade