Formalin 10%, Q Path®, buffered
Pot, 80 ml in a 150 ml plastic pot, 32
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.
Formaldehyde 4% is approximately equal to Formalin 10%.
Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Density:1.013 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-001-8
Documents
Certificates
Search by Lot NumberCutsheet VWRQ10.07040WH0002.pdf Cutsheet VWRQ10.300150WH01.pdf Cutsheet VWRQFOR0070AF59001.pdf
Technical Drawing
FOR0070AF59001
Images 360
Specifications
Specification Test Results
- Appearance:Colourless liquid
- Test formaldehyde:Passes test
- Density (20/4):1,012 - 1,014
- pH (20°C):6,8 - 7,2
Specifications
- Pack type:Pot
- Pk:32
- Pk Info:80 ml in a 150 ml plastic pot
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



