Order Entry
Italy
ContactUsLinkComponent
Human ST3GAL1 partial ORF ( NP_003024, 29 A.A. - 128 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00006482-Q012
Nome del fornitore:  Abnova
Human ST3GAL1 partial ORF ( NP_003024, 29 A.A. - 128 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00006482-Q012
Nome del fornitore:  Abnova
Frequently Bought Together