Order Entry
Italy
ContactUsLinkComponent
HOTTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BEEN PROVED NOT HAVING THE 3' - 5'EXONUCLEASE ACTIVITY. PRIOR THE FIRST PCR STEP THE HOTTAQ DNA POLYMERASE SHOULD BE ACTIVATED BY 15 MINUTE INCUBATION AT 95-97&DEG C. 1 * 2.500 EU
Catalog # ABNOD0011-3
Supplier:  Abnova
HOTTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BEEN PROVED NOT HAVING THE 3' - 5'EXONUCLEASE ACTIVITY. PRIOR THE FIRST PCR STEP THE HOTTAQ DNA POLYMERASE SHOULD BE ACTIVATED BY 15 MINUTE INCUBATION AT 95-97&DEG C. 1 * 2.500 EU
Catalog # ABNOD0011-3
Supplier:  Abnova
Frequently Bought Together