Order Entry
Italy
Orders LinkContactUsLinkComponent
Bottles, water sampling
Bottles, water sampling
125 ml
Catalog # LPIT299148
Supplier:   LP ITALIANA
Bottles, water sampling
Catalog # LPIT299148
Supplier:  LP ITALIANA
Bottles, water sampling
Bottles, water sampling
125 ml
Price
746,00 €
Log in for availability
1 * 350 pezzi
Product Details & Documents
PP, translucent, square, with screw cap.
  • Tamper evident screw caps
  • Sterile
  • Moulded graduations
Bottles for microbiological analysis of chlorinated or non-chlorinated water. Sodium thiosulphate inhibits the effects of chlorine, bromide and ozone present in the water. Ideal for flat storage (fridges, etc).
Specifications
  • Capacity:
    125 ml
  • Seq:
    02