Order Entry
Italy
Orders LinkContactUsLinkComponent
Isolator cleaning tool
Isolator cleaning tool
Sterile Mini AlphaMop™ covers polyester replacement covers and pads: TX7114, for use with TX7101 and TX7104, 125 covers and 25 pads/case
Catalog # 141-1046
Supplier:   Texwipe
Isolator cleaning tool
Catalog # 141-1046
Supplier:  Texwipe
Isolator cleaning tool
Isolator cleaning tool
Sterile Mini AlphaMop™ covers polyester replacement covers and pads: TX7114, for use with TX7101 and TX7104, 125 covers and 25 pads/case
Price
513,79 €
Log in for availability
1 * 125 pezzi
Product Details & Documents
Isolator cleaning tool: Mop hardware, replacement head, replacement handle.
Specially engineered to work with isolators and other small areas, this easy to manoeuvre tool offers lightweight, one handed operation. With a low profile, swivel head, the isolator cleaning tool gets into 'hard to reach' places.
Engineered for quick cover changes during operation, the padded head allows for a consistent mop across different surfaces without losing cleaning power. The rounded edges clean the isolator corners and prevent scratches or damage to surfaces.
  • Rapid change system allows user to replace covers/pads frequently, adhering to strict cleaning protocols
  • Polyester pad conforms to surfaces
  • Gets into 'hard to reach' areas with its low profile, swivel head
  • 100% polyester covers provide the cleanest substrates available for cleaning isolators
  • Offers one handed operation with lightweight handles
Isolator cleaning covers:
Made from 100% polyester or 100% polyester microdenier fabric. This fabric provides enhanced particle lifting, streak-free cleaning, plus enhanced sorption for working with solutions.
Enhanced absorbency and quick-wicking performance
Increased lifting and capturing of particles as small as microbes Streak-free cleaning.
Made from 100% polyester or 100% polyester microdenier fabric. This fabric provides enhanced particle lifting, streak-free cleaning, plus enhanced sorption for working with solutions.
Specifications
  • Description:
    Sterile Mini AlphaMop™ covers polyester replacement covers and pads: TX7114, for use with TX7101 and TX7104
  • Seq:
    7
  • Packed:
    125 covers and 25 pads/case