Order Entry
Italy
Orders LinkContactUsLinkComponent
Paraformaldehyde ≥97%

Paraformaldehyde ≥97%

Glass bottle for solids, 100 kg
Catalog # A11313.PG
Corrosive
Flammable
Health
Irritant
Paraformaldehyde ≥97%
Catalog # A11313.PG
CAS Number:  30525-89-4
Paraformaldehyde ≥97%
Paraformaldehyde ≥97%

Glass bottle for solids, 100 kg

CAS Number:  30525-89-4
Price
Inquire For Price
Log in for availability
1 * 100 kg
Product Details & Documents
Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • Merck Index:14,7025
  • Boiling Pt:135…136 °C (13 mmHg)
  • Melting Pt:120…170 °C (dec.)
  • Density:1.30
  • Flash Pt:71 °C (159 °F)
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2213
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.