Order Entry
Italy
ContactUsLinkComponent
Each v-21+k wear metals standard 50 µg/g, hydrocarbon oil, 400 g includes the following elements: ag, al, ba, b, cd, ca, cr, cu, fe, k, mg, mn, mo, na, ni, p, pb, si, sn, ti, v, zn. concentration: 50 µg/g, matrix: hydrocarbon oil and volume: 400 g. t 1 * 1 items
Catalog # PERCN9308372
Nome del fornitore:  PerkinElmer
Each v-21+k wear metals standard 50 µg/g, hydrocarbon oil, 400 g includes the following elements: ag, al, ba, b, cd, ca, cr, cu, fe, k, mg, mn, mo, na, ni, p, pb, si, sn, ti, v, zn. concentration: 50 µg/g, matrix: hydrocarbon oil and volume: 400 g. t 1 * 1 items
Catalog # PERCN9308372
Nome del fornitore:  PerkinElmer