Perkinelmer offers a wide selection of wear metal metallo-organic standards including v-21 wear metal standards concentration 10 µg/ g -400g. matrix: hydrocarbon, oilsize: 400 g. its features and benefits include - metallo-organic metals in hydrocarb 1 * 1 items
Catalog # PERCN9308324
Supplier: PerkinElmer